0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
DACOMITINIB (FREE BASE) 1 * 25 mg
ENZOENZCHM2210025
:
Enzo Life Sciences
DACOMITINIB (FREE BASE) 1 * 25 mg
ENZOENZCHM2210025
:
Enzo Life Sciences
Frequently Bought Together
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
: Merck
di-Sodium tetraborate decahydrate ACS, ISO, Reag. Ph. Eur. analytical reagent, Supelco®
: Merck
Test strips for peroxide, MQuant®, Supelco®
: Merck
Test strips for peracetic acid, MQuant®, Supelco®
: Merck
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Test kits, surfactants, Spectroquant®, Supelco®
: Merck
Test strips for free chlorine, MQuant®, Supelco®
: Merck
VWR®, Powder packs for determination of chlorine, reagents for photometers
: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
: Merck
Test kits, photometric, COD, LCK cuvette tests
: Hach