ContactUsLinkComponent
14 components: 100mg/l each of Bisphenol A [CAS:80-05-7] Bisphenol B [CAS:77-40-7] Bis(4-hydroxyphenyl)methane [CAS:620-92-8] Bisphenol S [CAS:80-09-1] a,a'-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene [CAS:2167-51-3] Bisphenol TMC [CAS:1291 1 * 5 Ampoul
Catalog # CMPLRD1044661.5
14 components: 100mg/l each of Bisphenol A [CAS:80-05-7] Bisphenol B [CAS:77-40-7] Bis(4-hydroxyphenyl)methane [CAS:620-92-8] Bisphenol S [CAS:80-09-1] a,a'-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene [CAS:2167-51-3] Bisphenol TMC [CAS:1291 1 * 5 Ampoul
Catalog # CMPLRD1044661.5
Frequently Bought Together