U-plan fluorite 40x objective 1 * 1 items
Katalogové číslo ECHLCMAG0405
Supplier: DISCOVER ECHO
Frequently Bought Together
Short-term tubes for accuro®
Supplier: Dräger
VWR® Counter-Pen™ Cell Counter
Supplier: VWR Collection
OIL IMPACTOR 0.1-1.0 MG/M3 1 * 10 items
Supplier: Dräger
VWR® pHenomenal® MU 6100 L Multi-Parameter Meter (pH/Conductivity/DO), Bench
Supplier: VWR Collection
VWR® HCO 304, Conductivity Meter, Handheld
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
Accessories for ThermoMixer™ F1,5 and FP
Supplier: EPPENDORF
VWR® VisiScope® STB150, Stereo Microscope
Supplier: VWR Collection
Conductivity meters, handheld, Cond 3110 / 3310
Supplier: WTW
Guillotine tubing cutter
Supplier: VICI JOUR RESEARCH
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
pH/mV/°C meters, bench, FiveEasy™ F20 / FiveEasy™ Plus FP20
Supplier: Mettler - Toledo
Plastic cuvette for LICO
Supplier: Hach
Density meters, Densito and DensitoPro
Supplier: Mettler - Toledo
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
GLASS CYLINDER FOR 10 ML BURETTE 1 * 1 items
Supplier: Mettler - Toledo