ContactUsLinkComponent
,,8 components: Dextran 40 [CAS:9004-54-0] 13g/l Maltotriose [CAS:1109-28-0] 14g/l D-(+)-Maltose Monohydrate [CAS:6363-53-7] 13g/l D-(+)-Glucose [CAS:50-99-7] 13g/l L-(+)-Lactic acid [CAS:79-33-4] 2g/l Glycerol [CAS:56-81-5] 2g/l Acetic a 1 * 100 mL
Catalog # CMPLRD094793L1
,,8 components: Dextran 40 [CAS:9004-54-0] 13g/l Maltotriose [CAS:1109-28-0] 14g/l D-(+)-Maltose Monohydrate [CAS:6363-53-7] 13g/l D-(+)-Glucose [CAS:50-99-7] 13g/l L-(+)-Lactic acid [CAS:79-33-4] 2g/l Glycerol [CAS:56-81-5] 2g/l Acetic a 1 * 100 mL
Catalog # CMPLRD094793L1
Frequently Bought Together