Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
5 COMPONENTS MIX 1 * 100 mL
Catalog # CMPLRM107014L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
5 COMPONENTS MIX 1 * 100 mL
Catalog # CMPLRM107014L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Electrode solutions and reagents for ion selective electrodes
Supplier: Hach
VWR® Powder Packs for Determination of Iron, Reagents for Photometers
Supplier: VWR Chemicals
NANOCOLOR® Photometric Tube Tests, Chromium
Supplier: MACHEREY-NAGEL
Sodium hydroxide 98%, flakes, pure
Supplier: Thermo Fisher Scientific