ContactUsLinkComponent
,,16 components: 100mg/l each of Al As Ba Cd Cr Cu Fe Mo Ni Pb Sb Se Sn Zn Mn P in HNO3 5% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM106271L1
,,16 components: 100mg/l each of Al As Ba Cd Cr Cu Fe Mo Ni Pb Sb Se Sn Zn Mn P in HNO3 5% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM106271L1