Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
L-Prolinamide 97%

L-Prolinamide 97%

Bottle, 25 g
Catalog # APOSOR3107-25G
Irritant
Supplier:   Apollo Scientific
L-Prolinamide 97%
Catalog # APOSOR3107-25G
Supplier:  Apollo Scientific
CAS Number:  7531-52-4
L-Prolinamide 97%

Bottle, 25 g

CAS Number:  7531-52-4
Price
1 850,00 Kč
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  H-Pro-NH₂, (S)-2-Pyrrolidinecarboxamide, H-Pro-NH2
  • CAS Number:7531-52-4
  • Formula:C₅H₁₀N₂O
  • MDL Number:MFCD00005253
  • Molecular Weight:114.15 g/mol
  • Melting Pt:95…97 °C
  • Storage Temperature:Ambient
  • EINECS:231-397-0
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications