Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
L-Prolinamide 97%

L-Prolinamide 97%

Bottle, 5 g
Catalog # APOSOR3107-5G
Irritant
Supplier:   Apollo Scientific
L-Prolinamide 97%
Catalog # APOSOR3107-5G
Supplier:  Apollo Scientific
CAS Number:  7531-52-4
L-Prolinamide 97%

Bottle, 5 g

CAS Number:  7531-52-4
Price
1 440,00 Kč
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  H-Pro-NH₂, (S)-2-Pyrrolidinecarboxamide, H-Pro-NH2
  • CAS Number:7531-52-4
  • Formula:C₅H₁₀N₂O
  • MDL Number:MFCD00005253
  • Molecular Weight:114.15 g/mol
  • Melting Pt:95…97 °C
  • Storage Temperature:Ambient
  • EINECS:231-397-0
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.