ContactUsLinkComponent
4x HF.E.35324 socket strip french / belgian version 4x HF.I.35305 perforated shelf, powder-coated sheet steel, RAL 7035 light grey, load capacity=75 kg 1x EP.ML.34775 bottom collecting sump standard, powder-coated sheet steel, RAL 7035 light grey, ca 1 * 1 items
Catalog # MSGA41103
Supplier:  asecos
4x HF.E.35324 socket strip french / belgian version 4x HF.I.35305 perforated shelf, powder-coated sheet steel, RAL 7035 light grey, load capacity=75 kg 1x EP.ML.34775 bottom collecting sump standard, powder-coated sheet steel, RAL 7035 light grey, ca 1 * 1 items
Catalog # MSGA41103
Supplier:  asecos
Frequently Bought Together