ContactUsLinkComponent
9 components: 10mg/l each of Hg Tl Ag Se Mo Bi Y In Ce in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 100 mL
Catalog # CMPLRM105519L1
9 components: 10mg/l each of Hg Tl Ag Se Mo Bi Y In Ce in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 100 mL
Catalog # CMPLRM105519L1
Frequently Bought Together