ContactUsLinkComponent
22 components: 100mg/l each of As Cd Co Cr Cu Fe Mn Mo Ni Pb Sb Se Sr Ti V Zn Al Ba Bi B Sn Zr in HNO3 5% CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temp 1 * 2 Ampoul
Catalog # CMPLRM086038L1.2
22 components: 100mg/l each of As Cd Co Cr Cu Fe Mn Mo Ni Pb Sb Se Sr Ti V Zn Al Ba Bi B Sn Zr in HNO3 5% CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temp 1 * 2 Ampoul
Catalog # CMPLRM086038L1.2