CHLOROFORM-D 1 * 10 g
Catalog # PROOTRCC366492-10
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Redox (ORP) Standards
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR® PESTINORM®, Gas Chromatography (GC) Certified Reference Material
Supplier: VWR Chemicals