ContactUsLinkComponent
Set 4 x 50ml, 23 components: 100mg/l each of Al Ba Bi Ca Ce Cr Co Cu Ga Fe In K Li Mg Mn Na Ni Pb Re Sr V Y Zn in HNO3 7% 1 * 200 mL
Catalog # CMPLRM105154L05.4
Set 4 x 50ml, 23 components: 100mg/l each of Al Ba Bi Ca Ce Cr Co Cu Ga Fe In K Li Mg Mn Na Ni Pb Re Sr V Y Zn in HNO3 7% 1 * 200 mL
Catalog # CMPLRM105154L05.4
Frequently Bought Together