ContactUsLinkComponent
Set 4 x 50ml, 14 components:100mg/l each of Sb Ge Hf Ir Mo Nb Pt Ru Ta Te Sn Ti W Zr in HCl 10% HF 1.2% 1 * 200 mL
Catalog # CMPLRM099566L05.4
Set 4 x 50ml, 14 components:100mg/l each of Sb Ge Hf Ir Mo Nb Pt Ru Ta Te Sn Ti W Zr in HCl 10% HF 1.2% 1 * 200 mL
Catalog # CMPLRM099566L05.4
Frequently Bought Together