ContactUsLinkComponent
Support frame, powder-coated steel, RAL 7035 light grey for hazardous materials work stations with width 900 mm with depth 600 mm, for model(s): GAP Version: ST version - standing (adjustable +30/-10 mm) dimensions W x D x H (mm): exterior: 900 x 600 1 * 1 items
Catalog # MSGA41054
Supplier:  asecos
Support frame, powder-coated steel, RAL 7035 light grey for hazardous materials work stations with width 900 mm with depth 600 mm, for model(s): GAP Version: ST version - standing (adjustable +30/-10 mm) dimensions W x D x H (mm): exterior: 900 x 600 1 * 1 items
Catalog # MSGA41054
Supplier:  asecos
Frequently Bought Together