Ethyl-tert-butylether (ETBE) CAS:637-92-3 EC:211-309-7 1 * 100 mg
Catalog # CMPLSB472.100MG
Supplier: CUSTOM MADE CHEMICALS LAB
Ethyl-tert-butylether (ETBE) CAS:637-92-3 EC:211-309-7 1 * 100 mg
Catalog # CMPLSB472.100MG
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Acetic acid 99% ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test strips for nickel, MQuant®, Supelco®
Supplier: Merck
Giemsa's stain solution for microscopy, Sigma-Aldrich®
Supplier: Merck
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Calcium acetate xhydrate, Supelco®
Supplier: Merck
Test strips, peracetic acid, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Test strips for iron, MQuant®, Supelco®
Supplier: Merck
Reference substances for GC analysis, Supelco®
Supplier: Merck
Sodium hydroxide 32% in aqueous solution
Supplier: Thermo Fisher Scientific
Sodium dodecyl sulphate (SDS) 0.01 mol/l in aqueous solution
Supplier: VWR Chemicals