ContactUsLinkComponent
6 components: 10ppb each of Li Mg Y Ce Tl Co in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 8 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 100 mL
  CMPLRM033114L1
6 components: 10ppb each of Li Mg Y Ce Tl Co in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 8 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 100 mL
  CMPLRM033114L1
Frequently Bought Together