ContactUsLinkComponent
Y-27632 is selective and potent ROCK inhibitor. Y-27632 plays various roles including contraction of vascular smooth muscle cells vi a ROCK signaling pathways. Furthermore, it suppresses the cell death of human embryonic stem cells (hESCs) and human 1 * 25 mg
Catalog # WAKF258-00703
Y-27632 is selective and potent ROCK inhibitor. Y-27632 plays various roles including contraction of vascular smooth muscle cells vi a ROCK signaling pathways. Furthermore, it suppresses the cell death of human embryonic stem cells (hESCs) and human 1 * 25 mg
Catalog # WAKF258-00703
Frequently Bought Together