ContactUsLinkComponent
The MassivEV™ EV Purification Column PS and MassivEV™ Purification Buffer Set have been specifically designed for the isolation and purification of extracellular vesicles (EVs), including exosomes, from large volumes of cell culture supernatants. Thi 1 * 5 mL
Catalog # WAKF137-19493
The MassivEV™ EV Purification Column PS and MassivEV™ Purification Buffer Set have been specifically designed for the isolation and purification of extracellular vesicles (EVs), including exosomes, from large volumes of cell culture supernatants. Thi 1 * 5 mL
Catalog # WAKF137-19493
Frequently Bought Together