BOTTLE W/N AMBER 1000ML GL68 1 * 18 items
Catalog # 215-4706
Supplier: VWR Collection
Frequently Bought Together
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Samplers for liquids, TeleScoop
Supplier: BURKLE
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Screw Caps for Wide Neck Bottles
Supplier: RIXIUS
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF