Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KLERCIDE LOW RESIDUE QUAT 5L WFI STERILE 1 * 4 KS
Catalog # SHIE3084140
Supplier:
ECOLAB CONTAMINATION CONTROL
Certificates
Expand image
KLERCIDE LOW RESIDUE QUAT 5L WFI STERILE 1 * 4 KS
Catalog # SHIE3084140
Supplier:
ECOLAB CONTAMINATION CONTROL
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran stabilised, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
o-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
N,O-Bis(trimethylsilyl)acetamide : chlorotrimethylsilane : 1-(trimethylsilyl)imidazole (BSA+TMCS+TMSI) silylating mixture II according to Horning, LiChropur™ for GC derivatisation, Supelco®
Supplier: Merck
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals