2 components: 100mg/l each of Chlordecone hydrate [CAS:143-50-0] Mirex [CAS:2385-85-5] in Acetone CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a freezer at -18C or below 1 * 5 Ampoul
Catalog # CMPLRD1026431.5
Dodavatel: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together