ContactUsLinkComponent
DNA (-) Control CISH Probe is designed to be used for the qualitative detection of unspecific background staining in formalin-fixed, paraffin-embedded tissue or cells by chromogenic in situ hybridization (CISH). 1 * 100 µl
Catalog # ABNOCO0007.100UL
Supplier:  Abnova
DNA (-) Control CISH Probe is designed to be used for the qualitative detection of unspecific background staining in formalin-fixed, paraffin-embedded tissue or cells by chromogenic in situ hybridization (CISH). 1 * 100 µl
Catalog # ABNOCO0007.100UL
Supplier:  Abnova
Frequently Bought Together