ContactUsLinkComponent
The Heat Pretreatment Solution (EDTA) is intended to be used to heat pretreatment of formalin-fixed, paraffin-embedded specimens in CISH. 1 * 500 mL
Catalog # ABNOU0542.500ML
Supplier:  Abnova
The Heat Pretreatment Solution (EDTA) is intended to be used to heat pretreatment of formalin-fixed, paraffin-embedded specimens in CISH. 1 * 500 mL
Catalog # ABNOU0542.500ML
Supplier:  Abnova
Frequently Bought Together