The Rabbit Anti-DIG is intended to be used for detection steps in CISH on formalin-fixed, paraffin-embedded specimens. 1 * 4 mL
Catalog # ABNOU0527.4ML
Supplier: Abnova
The Rabbit Anti-DIG is intended to be used for detection steps in CISH on formalin-fixed, paraffin-embedded specimens. 1 * 4 mL
Catalog # ABNOU0527.4ML
Supplier: Abnova
Frequently Bought Together
VWR®, Microcentrifuge Tubes and Screw Caps
Supplier: VWR Collection
Sampling spatula, Sterileware®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Cell culture flasks, Falcon®
Supplier: Corning
VWR® Drosophila Vials
Supplier: VWR Collection
Cryoboxes, 133×133 mm
Supplier: TENAK
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Microplates, 96-well
Supplier: Greiner Bio-One
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Ultra-High Performance, Centrifuge Tubes, Light-Sensitive
Supplier: VWR Collection
VWR® Cryoboxes and Dividers
Supplier: VWR Collection
Containers, wide mouth, with screw cap, Corning® Gosselin™
Supplier: Corning
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
Microcentrifuge tubes, 0,6/1,5/2,0 ml, graduated, with snap cap
Supplier: Thermo Fisher Scientific
Sampling scoops, PS, white, double-bagged, Sterileware®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Collagen I-coated microplates, Corning® BioCoat®
Supplier: Corning
Cryostorage boxes, KeepIT-100™, WHEATON®
Supplier: DWK Life Sciences