ContactUsLinkComponent
The Anti-Rabbit AP Polymer is intended to be used for detection steps in CISH on formalin-fixed, paraffin-embedded specimens. 1 * 4 mL
Catalog # ABNOU0528.4ML
Supplier:  Abnova
The Anti-Rabbit AP Polymer is intended to be used for detection steps in CISH on formalin-fixed, paraffin-embedded specimens. 1 * 4 mL
Catalog # ABNOU0528.4ML
Supplier:  Abnova
Frequently Bought Together