0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
The Mayer's Hematoxylin Solution is intended to be used for counterstaining steps in CISH on formalin-fixed, paraffin-embedded speci mens. 1 * 20 mL
ABNOU0531.20ML
:
Abnova
The Mayer's Hematoxylin Solution is intended to be used for counterstaining steps in CISH on formalin-fixed, paraffin-embedded speci mens. 1 * 20 mL
ABNOU0531.20ML
:
Abnova
Frequently Bought Together
96-Well clear polystyrene microplates, Corning®
: Corning
VWR® Blender Bags
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Reagent/centrifuge tube with screw cap
: Greiner Bio-One
VWR®, Ice Pans, with or without Lid
: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
Cryoboxes, 133×133 mm
: TENAK
Culture media bottles, square, sterile, Nalgene®
: Thermo Fisher Scientific
VWR® Standard Line Transfer Pipettes
: VWR Collection
Cryogenic tubes, CryoTubes™, Nunc™
: Thermo Fisher Scientific
VWR®, Sampling Spoons
: VWR Collection
VWR®, Cell Strainers
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Cryoboxes, B69/B50/B80/B99-Series
: GLW
Centrifuge tubes, Falcon®
: Corning