ContactUsLinkComponent
The Nuclear Red Solution is intended to be used for counterstaining steps in CISH on formalin-fixed, paraffin-embedded specimens. 1 * 20 mL
Catalog # ABNOU0533.20ML
Supplier:  Abnova
The Nuclear Red Solution is intended to be used for counterstaining steps in CISH on formalin-fixed, paraffin-embedded specimens. 1 * 20 mL
Catalog # ABNOU0533.20ML
Supplier:  Abnova
Frequently Bought Together