ContactUsLinkComponent
The Fixogum Rubber Cement is a glue used to seal the samples on a slide with a coverslip in FISH to protect the sample from drying o ut during hybridization. 1 * 125 g
Catalog # ABNOU0535.125G
Supplier:  Abnova
The Fixogum Rubber Cement is a glue used to seal the samples on a slide with a coverslip in FISH to protect the sample from drying o ut during hybridization. 1 * 125 g
Catalog # ABNOU0535.125G
Supplier:  Abnova
Frequently Bought Together