ContactUsLinkComponent
The Fixogum Rubber Cement is a glue used to seal the samples on a slide with a coverslip in FISH to protect the sample from drying o ut during hybridization. 1 * 50 g
Catalog # ABNOU0536.50G
Supplier:  Abnova
The Fixogum Rubber Cement is a glue used to seal the samples on a slide with a coverslip in FISH to protect the sample from drying o ut during hybridization. 1 * 50 g
Catalog # ABNOU0536.50G
Supplier:  Abnova
Frequently Bought Together