ContactUsLinkComponent
MDM2/CEN 12 Probe is designed for the qualitative detection of human MDM2 gene amplifications as well as the detection of chromosome 12 alpha satellites in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH 1 * 100 µl
Catalog # ABNOCG0038.100UL
Supplier:  Abnova
MDM2/CEN 12 Probe is designed for the qualitative detection of human MDM2 gene amplifications as well as the detection of chromosome 12 alpha satellites in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH 1 * 100 µl
Catalog # ABNOCG0038.100UL
Supplier:  Abnova
Frequently Bought Together