ContactUsLinkComponent
ERBB2/D17S122 Probe is designed for the qualitative detection of human ERBB2 gene amplifications as well as the detection of the D17 S122 locus in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH). 1 * 100 µl
Catalog # ABNOCG0040.100UL
Supplier:  Abnova
ERBB2/D17S122 Probe is designed for the qualitative detection of human ERBB2 gene amplifications as well as the detection of the D17 S122 locus in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH). 1 * 100 µl
Catalog # ABNOCG0040.100UL
Supplier:  Abnova
Frequently Bought Together