ContactUsLinkComponent
NTRK3 Split Probe is designed for the qualitative detection of translocations involving the human NTRK3 gene at 15q25.3 in formalin- fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH). 1 * 100 µl
Catalog # ABNOCS0021.100UL
Supplier:  Abnova
NTRK3 Split Probe is designed for the qualitative detection of translocations involving the human NTRK3 gene at 15q25.3 in formalin- fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH). 1 * 100 µl
Catalog # ABNOCS0021.100UL
Supplier:  Abnova
Frequently Bought Together