ContactUsLinkComponent
ERBB2/CEN 17 CISH Probe Kit is designed for the qualitative detection of amplifications involving the human ERBB2 gene as well as th e detection of chromosome 17 alpha satellites in formalin-fixed, paraffin-embedded specimens by chromogenic in sit 1 * 1 KIT
Catalog # ABNOKA6901.1KIT
Supplier:  Abnova
ERBB2/CEN 17 CISH Probe Kit is designed for the qualitative detection of amplifications involving the human ERBB2 gene as well as th e detection of chromosome 17 alpha satellites in formalin-fixed, paraffin-embedded specimens by chromogenic in sit 1 * 1 KIT
Catalog # ABNOKA6901.1KIT
Supplier:  Abnova
Frequently Bought Together