ContactUsLinkComponent
PEG Blocking Buffer for ELISA contain polyoxyethylene detergents that interfere with PEG and PEG antibody detection. PEGB125-1 effec tively blocks and stabilizes PEG-protein and anti-PEG coated ELISA plates if used as directed. 1 * 125 mL
Catalog # ABNOU0521.125ML
Supplier:  Abnova
PEG Blocking Buffer for ELISA contain polyoxyethylene detergents that interfere with PEG and PEG antibody detection. PEGB125-1 effec tively blocks and stabilizes PEG-protein and anti-PEG coated ELISA plates if used as directed. 1 * 125 mL
Catalog # ABNOU0521.125ML
Supplier:  Abnova
Frequently Bought Together