ContactUsLinkComponent
3-OH AG(cap1) is used to form a Cap 1 structure by capping the 5' end of the mRNA during in vitro transcription. 1 * 50 µl
Catalog # ABNOU0451.50UL
Supplier:  Abnova
3-OH AG(cap1) is used to form a Cap 1 structure by capping the 5' end of the mRNA during in vitro transcription. 1 * 50 µl
Catalog # ABNOU0451.50UL
Supplier:  Abnova
Frequently Bought Together