ContactUsLinkComponent
RNase III is ribonuclease III (RNase III) recombinantly expressed by E. coli. This specific exonuclease cleaves double-stranded RNA (dsRNA) and generates 12-35bp dsRNA fragments with 5'-PO4 and 3'-OH, 3' overhangs. 1 * 125 µl
Catalog # ABNOP8872.125UL
Supplier:  Abnova
RNase III is ribonuclease III (RNase III) recombinantly expressed by E. coli. This specific exonuclease cleaves double-stranded RNA (dsRNA) and generates 12-35bp dsRNA fragments with 5'-PO4 and 3'-OH, 3' overhangs. 1 * 125 µl
Catalog # ABNOP8872.125UL
Supplier:  Abnova
Frequently Bought Together