Inorganic pyrophosphate (PPi) is generated as a reaction byproduct in many biosynthetic reactions which utilize ATP, including in vi tro transcription and DNA polymerization. Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of inorganic pyr 1 * 10 Unit
Catalog # ABNOP8871.10UNITS
Supplier: Abnova
Frequently Bought Together