Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Stool DNA Purification Kit (Pre-filled) is designed for the optimal automated extraction of DNA from stool of human and other specie s. 1 * 4 KIT
Catalog # ABNOU0433.4KITS
Supplier:
Abnova
Expand image
Stool DNA Purification Kit (Pre-filled) is designed for the optimal automated extraction of DNA from stool of human and other specie s. 1 * 4 KIT
Catalog # ABNOU0433.4KITS
Supplier:
Abnova
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Blender Bags
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Transfer pipettes
Supplier: RATIOLAB
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection