0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Labeled FISH probes for identification of gene translocation using Fluorescent In Situ Hybridization Technique. (Technology) 1 * 100 µl
ABNOFG0257.100UL
:
Abnova
Labeled FISH probes for identification of gene translocation using Fluorescent In Situ Hybridization Technique. (Technology) 1 * 100 µl
ABNOFG0257.100UL
:
Abnova
Frequently Bought Together
Reagent/centrifuge tube with screw cap
: Greiner Bio-One
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Conical Sample Containers, with Snap-On Lids
: VWR Collection
VWR® PCR Tubes, 0,2 ml
: VWR Collection