Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Labeled FISH probes for identification of gene amplification using Fluorescent In Situ Hybridization Technique. (Technology) 1 * 100 µl
Catalog # ABNOFG0261.100UL
Supplier:
Abnova
Expand image
Labeled FISH probes for identification of gene amplification using Fluorescent In Situ Hybridization Technique. (Technology) 1 * 100 µl
Catalog # ABNOFG0261.100UL
Supplier:
Abnova
Frequently Bought Together
VWR®, Breatheable Rayon Film Seals for Biological Cultures
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
Cryoboxes, 133×133 mm
Supplier: TENAK
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Multiwell cell culture plates, Falcon®
Supplier: Corning
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection