Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Labeled FISH probes for identification of gene split using Fluorescent In Situ Hybridization Technique. (Technology) 1 * 100 µl
Catalog # ABNOFS0106.100UL
Supplier:
Abnova
Expand image
Labeled FISH probes for identification of gene split using Fluorescent In Situ Hybridization Technique. (Technology) 1 * 100 µl
Catalog # ABNOFS0106.100UL
Supplier:
Abnova
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection