Labeled FISH probes for identification of gene split using Fluorescent In Situ Hybridization Technique. (Technology) 1 * 100 µl
Catalog # ABNOFS0109.100UL
Supplier: Abnova
Labeled FISH probes for identification of gene split using Fluorescent In Situ Hybridization Technique. (Technology) 1 * 100 µl
Catalog # ABNOFS0109.100UL
Supplier: Abnova
Frequently Bought Together
Centrifuge tubes, 15 ml, Corning®
Supplier: Corning
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Multiwell cell culture plates, Falcon®
Supplier: Corning
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Half Size, Cardboard
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection