Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ALDH Activity Assay Kit uses a fluorescence-based method for detecting ALDH activity in tissue homogenates, cell culture samples, an d purified ALDH preparations. 1 * 1 KIT
Catalog # ABNOKA6687.1KIT
Supplier:
Abnova
Expand image
ALDH Activity Assay Kit uses a fluorescence-based method for detecting ALDH activity in tissue homogenates, cell culture samples, an d purified ALDH preparations. 1 * 1 KIT
Catalog # ABNOKA6687.1KIT
Supplier:
Abnova
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Cryoboxes with Dividers
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, Sterile
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection