ContactUsLinkComponent
Epstein-Barr Virus IgA Rapid Test is for qualitive and semi-quantitative of EBV infection using IgA antibodies in human serum or pl asma. 1 * 1 KIT
Catalog # ABNOKA6654.1KIT
Supplier:  Abnova
Epstein-Barr Virus IgA Rapid Test is for qualitive and semi-quantitative of EBV infection using IgA antibodies in human serum or pl asma. 1 * 1 KIT
Catalog # ABNOKA6654.1KIT
Supplier:  Abnova
Frequently Bought Together