Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Epstein-Barr Virus IgM Rapid Test is for qualitive and semi-quantitative of EBV infection using IgM antibodies in human serum or pl asma. 1 * 1 KIT
Catalog # ABNOKA6656.1KIT
Supplier:
Abnova
Expand image
Epstein-Barr Virus IgM Rapid Test is for qualitive and semi-quantitative of EBV infection using IgM antibodies in human serum or pl asma. 1 * 1 KIT
Catalog # ABNOKA6656.1KIT
Supplier:
Abnova
Frequently Bought Together
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Test tubes
Supplier: Greiner Bio-One
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Centrifuge tubes, 15 ml, Corning®
Supplier: Corning
Eppendorf Tubes® Microtubes with Screw Cap, 5,0 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection