ContactUsLinkComponent
GAM Broth contains haemin from digested serum, it is used to cultivate anaerobic bacteria. It can also be used to detect other microorganisms such as streptococci, pneumococci and meningococci and is suitable for blood culture 1 * 1 items
Catalog # NISS05422-GAB-0300
GAM Broth contains haemin from digested serum, it is used to cultivate anaerobic bacteria. It can also be used to detect other microorganisms such as streptococci, pneumococci and meningococci and is suitable for blood culture 1 * 1 items
Catalog # NISS05422-GAB-0300
Frequently Bought Together