ContactUsLinkComponent
Initial Calibration Verification Standard - 26 components: Fe 1000mg/l K 1000mg/l Ca 1000mg/l Na 1000mg/l Mg 1000mg/l Sr 100mg/l Ag 10mg/l Al 10mg/l As 10mg/l Ba 10mg/l Be 10mg/l Cd 10mg/l Co 10mg/l Cr 10mg/l Cu 10mg/l M 1 * 100 mL
Catalog # CMPL5183-4682.L1
Initial Calibration Verification Standard - 26 components: Fe 1000mg/l K 1000mg/l Ca 1000mg/l Na 1000mg/l Mg 1000mg/l Sr 100mg/l Ag 10mg/l Al 10mg/l As 10mg/l Ba 10mg/l Be 10mg/l Cd 10mg/l Co 10mg/l Cr 10mg/l Cu 10mg/l M 1 * 100 mL
Catalog # CMPL5183-4682.L1