8 components: P 2500mg/l B 500mg/l Rb 500mg/l Si 500mg/l As 2.5mg/l Sn 2.5mg/l Pb 2.5mg/l Ti 0.5mg/l in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 8 months. (4 months after opening) Store under normal laboratory conditions, at te 1 * 5 Ampoul
Catalog # CMPLRM062418L1.5
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together