ContactUsLinkComponent
7 components: K 20000mg/l Ca 2500mg/l Al 20mg/l Cu 20mg/l Sr 20mg/l Ba 4mg/l Ag 2mg/l in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 12 months. (6 months after opening) Store under normal laboratory conditions, at temperatures betwe 1 * 5 Ampoul
Catalog # CMPLRM069206L1.5
7 components: K 20000mg/l Ca 2500mg/l Al 20mg/l Cu 20mg/l Sr 20mg/l Ba 4mg/l Ag 2mg/l in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 12 months. (6 months after opening) Store under normal laboratory conditions, at temperatures betwe 1 * 5 Ampoul
Catalog # CMPLRM069206L1.5
Frequently Bought Together